Please login to your online account to display your discounted pricing

anti-CD1a, Clone: C1A/711, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2330181ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2330181M

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Recombinant human CD1a protein
Isotype κ
Quantity 1mL
Format Purified
Host Species Mouse
Gene Accession No. P06126
Clone C1A/711
Primary or Secondary Primary
Concentration 0.2mg/mL
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 909
Formulation PBS with 0.05% BSA.
Cross Reactivity Human
Regulatory Status RUO
Antigen CD1a