Please login to your online account to display your discounted pricing

anti-CD15/Lewis X, Clone: published as MMA and Leu-M1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP229410

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP229410

  • / Each
 

Description & Specifications

Specifications

Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Isotype κ
Quantity 0.5mL
Antigen CD15/Lewis X
Immunogen The U937 histiocytic cell line was used as the immunogen for the CD15 Leu-M1 antibody.
Gene ID (Entrez) 2526
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation Culture Medium
Conjugate Unlabeled
Gene Accession No. P22083
Test Specificity CD15 plays a role in mediating phagocytosis, bactericidal activity, and chemotaxis. It is present on less than 95% of granulocytes including neutrophils and eosinophils and to a lesser degree on monocytes. In addition, CD15 is expressed in Reed-Sternberg cells and some epithelial cells. CD15 antibody is very useful in the identification of Hodgkin's disease. CD15 is occasionally expressed in large cell lymphomas of both B and T phenotypes which otherwise have a quite distinct histological appearance.
Regulatory Status RUO
Cross Reactivity Human
Clone published as MMA and Leu-M1
Format Concentrated