Please login to your online account to display your discounted pricing

anti-CD15/Lewis X, Clone: published as MMA and Leu-M1, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone published as MMA and Leu-M1
Host Species Mouse
Isotype κ
Immunogen The U937 histiocytic cell line was used as the immunogen for the CD15 Leu-M1 antibody.
Antigen CD15/Lewis X
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP229410

 
novus biologicals canada
NBP229410
Unlabeled 0.5mL Each for $321.00

NBP2294101M

 
novus biologicals canada
NBP2294101ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone published as MMA and Leu-M1
Host Species Mouse
Isotype κ
Immunogen The U937 histiocytic cell line was used as the immunogen for the CD15 Leu-M1 antibody.
Antigen CD15/Lewis X
Monoclonal or Polyclonal Monoclonal
Format Concentrated
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity CD15 plays a role in mediating phagocytosis, bactericidal activity, and chemotaxis. It is present on less than 95% of granulocytes including neutrophils and eosinophils and to a lesser degree on monocytes. In addition, CD15 is expressed in Reed-Sternberg cells and some epithelial cells. CD15 antibody is very useful in the identification of Hodgkin's disease. CD15 is occasionally expressed in large cell lymphomas of both B and T phenotypes which otherwise have a quite distinct histological appearance.
Gene Accession No. P22083
Gene ID (Entrez) 2526
Formulation Culture Medium