Please login to your online account to display your discounted pricing

anti-CD15/Lewis X, Clone: published as MMA and Leu-M1, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP2294101ML

 View all options for this product


For Research Use Only

Catalog No. NBP2294101M


Description & Specifications


Antigen CD15/Lewis X
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Clone published as MMA and Leu-M1
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Formulation Culture Medium
Gene Accession No. P22083
Host Species Mouse
Immunogen The U937 histiocytic cell line was used as the immunogen for the CD15 Leu-M1 antibody.
Isotype κ
Quantity 1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2526
Test Specificity CD15 plays a role in mediating phagocytosis, bactericidal activity, and chemotaxis. It is present on less than 95% of granulocytes including neutrophils and eosinophils and to a lesser degree on monocytes. In addition, CD15 is expressed in Reed-Sternberg cells and some epithelial cells. CD15 antibody is very useful in the identification of Hodgkin's disease. CD15 is occasionally expressed in large cell lymphomas of both B and T phenotypes which otherwise have a quite distinct histological appearance.