Please login to your online account to display your discounted pricing

anti-CD14, Clone: RPA-M1, Novus Biologicals™

$109.00 - $329.00

Specifications

Clone RPA-M1
Host Species Mouse
Isotype κ
Immunogen Human PHA-stimulated PBMC were used as immunogen.
Antigen CD14
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP225194

 
novus biologicals canada
NBP225194
Unlabeled 0.1mg Each for $329.00

NBP225194SS

 
novus biologicals canada
NBP225194SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Clone RPA-M1
Host Species Mouse
Isotype κ
Immunogen Human PHA-stimulated PBMC were used as immunogen.
Antigen CD14
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Test Specificity This product reacts with Human.
Gene Accession No. NP_000582
Gene ID (Entrez) 929
Formulation CD14 antibody at 0.5mg/mL in PBS containing 0.05% BSA
Concentration 0.5mg/mL