Please login to your online account to display your discounted pricing

anti-CD11c, Clone: HC1/1, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone HC1/1
Host Species Mouse
Isotype κ
Immunogen Human monocytic U937 phorbol ester-treated cells
Antigen CD11c
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2329241M

 
novus biologicals canada
NBP2329241ML
Unlabeled 1mL Each for $429.00

NBP232924

 
novus biologicals canada
NBP232924
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone HC1/1
Host Species Mouse
Isotype κ
Immunogen Human monocytic U937 phorbol ester-treated cells
Antigen CD11c
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Gene Accession No. P20702
Gene ID (Entrez) 3687
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL