Please login to your online account to display your discounted pricing

anti-CD11c, Clone: HC1/1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329241ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329241M

  • / Each
 

Description & Specifications

Specifications

Quantity 1mL
Isotype κ
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Cross Reactivity Human
Antigen CD11c
Format Purified
Clone HC1/1
Immunogen Human monocytic U937 phorbol ester-treated cells
Host Species Mouse
Regulatory Status RUO
Concentration 0.2mg/mL
Gene Accession No. P20702
Primary or Secondary Primary
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3687