missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Mouse CCL6/C10, Polyclonal, R&D Systems

$467.98 - $467.98

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. P27784
Gene ID (Entrez) 20305
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AF487 R & D Systems
RDSAF487
100μg Each for $467.98
AB487NA R & D Systems
RDSAB487NA
1mg Each for $467.98
Specifications

Description

Host Species: Goat
Species Reactivity:Mouse
Immunogen:E. coli-derived recombinant mouse CCL6/C10, Gly22-Ala116, Accession No. P27784

Applications:

Neutralization, Western Blotting

Description & Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
P27784
20305