Please login to your online account to display your discounted pricing

anti-Mouse CCL3/MIP-1 alpha, Polyclonal, R&D Systems


Manufacturer: r & d systems  RDSAB450NA

 View more versions of this product

Catalog No. AB450NA

  • / Each
 

Description & Specifications

Specifications

Isotype IgG
Quantity 1mg
Regulatory Status RUO
Immunogen E. coli-derived recombinant mouse CCL3/MIP-1α, Ala24-Ala92, Accession No. Q5QNW0
Cross Reactivity Mouse
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Antigen CCL3/MIP-1 alpha
Format Affinity Purified
Host Species Goat
Gene ID (Entrez) 20302
Gene Accession No. Q5QNW0
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.