missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-Mouse CCL3/MIP-1 alpha, Polyclonal, R&D Systems

$454.80 - $467.46

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. Q5QNW0
Gene ID (Entrez) 20302
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AB450NA R & D Systems
RDSAB450NA
1mg Each for $454.80
AF450NA R & D Systems
RDSAF450NA
100μg Each for $467.46
Specifications

Description

Host Species: Goat
Species Reactivity:Mouse
Immunogen:E. coli-derived recombinant mouse CCL3/MIP-1α, Ala24-Ala92, Accession No. Q5QNW0

Applications:

Neutralization, Western Blotting

Description & Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Q5QNW0
20302