Please login to your online account to display your discounted pricing

anti-CCL20/MIP-3 alpha, Clone: 3N2D9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227145

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227145

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Test Specificity This monoclonal antibody is specific to Human MIP-3 it will not cross-react with Mouse MIP-3.
Gene ID (Entrez) 6364
Applications Western Blotting
Antigen CCL20/MIP-3 alpha
Format Purified
Host Species Mouse
Regulatory Status RUO
Clone 3N2D9
Immunogen Full length recombinant human MIP-3 was used as immunogen for this antibody.
Quantity 0.1mg
Cross Reactivity Human
Concentration 0.5mg/mL
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal