Please login to your online account to display your discounted pricing

anti-CCL20/MIP-3 alpha, Clone: 3N2D9, Novus Biologicals™

$109.00 - $255.80

Specifications

Clone 3N2D9
Host Species Murine
Immunogen Full length recombinant human MIP-3 was used as immunogen for this antibody.
Antigen CCL20/MIP-3 alpha
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227145SS

 
novus biologicals canada
NBP227145SS
Unlabeled 0.025mg Each for $109.00

NBP227145

 
novus biologicals canada
NBP227145
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Clone 3N2D9
Host Species Murine
Immunogen Full length recombinant human MIP-3 was used as immunogen for this antibody.
Antigen CCL20/MIP-3 alpha
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity This monoclonal antibody is specific to Human MIP-3 it will not cross-react with Mouse MIP-3.
Gene ID (Entrez) 6364
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL