Please login to your online account to display your discounted pricing

anti-CCL2/MCP1, Clone: 2D8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP222115

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP222115

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Antigen CCL2/MCP1
Format Precipitation
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Isotype IgG1
Regulatory Status RUO
Gene Accession No. P13500
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Gene ID (Entrez) 6347
Concentration 1.0mg/mL
Conjugate Unlabeled
Formulation PBS
Quantity 0.1mL
Immunogen Purified recombinant fragment of human MCP1 expressed in &italic_begin;E. coli&italic_end;. [UniProt No. P13500]
Clone 2D8