Please login to your online account to display your discounted pricing

anti-CCL2/MCP1, Clone: 2D8, Novus Biologicals™

$164.00 - $384.00

Specifications

Clone 2D8
Host Species Murine
Isotype IgG1
Immunogen Purified recombinant fragment of human MCP1 expressed in &italic_begin;E. coli&italic_end;. [UniProt No. P13500]
Antigen CCL2/MCP1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP222115SS

 
novus biologicals canada
NBP222115SS
Unlabeled 0.025mL Each for $164.00

NBP222115

 
novus biologicals canada
NBP222115
Unlabeled 0.1mL Each for $384.00
Description & Specifications

Specifications

Clone 2D8
Host Species Murine
Isotype IgG1
Immunogen Purified recombinant fragment of human MCP1 expressed in &italic_begin;E. coli&italic_end;. [UniProt No. P13500]
Antigen CCL2/MCP1
Monoclonal or Polyclonal Monoclonal
Format Precipitation
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Accession No. P13500
Gene ID (Entrez) 6347
Formulation PBS
Concentration 1.0mg/mL