Please login to your online account to display your discounted pricing

anti-CCL2/MCP1, Clone: 2D8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP222115SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP222115SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Concentration 1.0mg/mL
Conjugate Unlabeled
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Purified recombinant fragment of human MCP1 expressed in &italic_begin;E. coli&italic_end;. [UniProt No. P13500]
Isotype IgG1
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Regulatory Status RUO
Format Precipitation
Host Species Mouse
Quantity 0.025mL
Clone 2D8
Antigen CCL2/MCP1
Formulation PBS
Gene Accession No. P13500
Gene ID (Entrez) 6347
Primary or Secondary Primary