Please login to your online account to display your discounted pricing

anti-CCL16/HCC-4/LEC, Clone: LEC66, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232857

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232857

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Immunogen Recombinant human HCC-4 protein
Format Purified
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Cross Reactivity Human
Formulation PBS with 0.05% BSA.
Gene Accession No. O15467
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Clone LEC66
Conjugate Unlabeled
Primary or Secondary Primary
Gene ID (Entrez) 6360
Concentration 0.2mg/mL
Antigen CCL16/HCC-4/LEC