Please login to your online account to display your discounted pricing

anti-CCL16/HCC-4/LEC, Clone: LEC66, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone LEC66
Host Species Mouse
Isotype κ
Immunogen Recombinant human HCC-4 protein
Antigen CCL16/HCC-4/LEC
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232857

 
novus biologicals canada
NBP232857
Unlabeled 0.5mL Each for $321.00

NBP2328571M

 
novus biologicals canada
NBP2328571ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone LEC66
Host Species Mouse
Isotype κ
Immunogen Recombinant human HCC-4 protein
Antigen CCL16/HCC-4/LEC
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Gene Accession No. O15467
Gene ID (Entrez) 6360
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL