Please login to your online account to display your discounted pricing

anti-CCL16/HCC-4/LEC, Clone: LEC66, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328571ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328571M

  • / Each
 

Description & Specifications

Specifications

Antigen CCL16/HCC-4/LEC
Applications ELISA
Clone LEC66
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. O15467
Host Species Mouse
Immunogen Recombinant human HCC-4 protein
Isotype κ
Quantity 1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6360