Please login to your online account to display your discounted pricing

anti-Caspase-8, Clone: FLICE 4-1-20, Novus Biologicals™


Manufacturer: novus biologicals canada  NB500208SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB500208SS

  • / Each
 

Description & Specifications

Specifications

Quantity 0.05mL
Format Ascites
Isotype κ
Regulatory Status RUO
Immunogen Full-length recombinant human Caspase 8 [UniProt No. Q14790]
Clone FLICE 4-1-20
Host Species Mouse
Gene Accession No. Q14790
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 841
Primary or Secondary Primary
Conjugate Unlabeled
Applications Western Blotting
Cross Reactivity Human
Antigen Caspase-8