Please login to your online account to display your discounted pricing

anti-Caspase-8, Clone: FLICE 4-1-20, Novus Biologicals™


Manufacturer: novus biologicals canada  NB500208

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB500208

  • / Each
 

Description & Specifications

Specifications

Antigen Caspase-8
Applications Western Blot
Clone FLICE 4-1-20
Conjugate Unlabeled
Cross Reactivity Human
Format Ascites
Gene Accession No. Q14790
Host Species Murine
Immunogen Full-length recombinant human Caspase 8 [UniProt No. Q14790]
Isotype κ
Quantity 0.2mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 841
Target Species Human