Please login to your online account to display your discounted pricing

anti-Caspase-8, Clone: FLICE 4-1-20, Novus Biologicals™

$109.00 - $259.02

Specifications

Clone FLICE 4-1-20
Host Species Murine
Isotype κ
Immunogen Full-length recombinant human Caspase 8 [UniProt No. Q14790]
Antigen Caspase-8
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB500208

 
novus biologicals canada
NB500208
Unlabeled 0.2mL Each for $259.02

NB500208SS

 
novus biologicals canada
NB500208SS
Unlabeled 0.05mL Each for $109.00
Description & Specifications

Specifications

Clone FLICE 4-1-20
Host Species Murine
Isotype κ
Immunogen Full-length recombinant human Caspase 8 [UniProt No. Q14790]
Antigen Caspase-8
Monoclonal or Polyclonal Monoclonal
Format Ascites
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Gene Accession No. Q14790
Gene ID (Entrez) 841