Please login to your online account to display your discounted pricing

anti-Caspase-7, Clone: 25B881.1, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 25B881.1
Host Species Mouse
Isotype IgG1
Immunogen This antibody was generated against recombinant protein of active form of human Caspase-7.
Antigen Caspase-7
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056529

 
novus biologicals canada
NB10056529
Unlabeled 0.1mg Each for $329.00

NB10056529T

 
novus biologicals canada
NB10056529SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 25B881.1
Host Species Mouse
Isotype IgG1
Immunogen This antibody was generated against recombinant protein of active form of human Caspase-7.
Antigen Caspase-7
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity In Jurkat, a 35 kDa band is observed.
Gene Accession No. P55210
Gene ID (Entrez) 840
Formulation PBS containing 0.2% gelatin