Please login to your online account to display your discounted pricing

anti-Caspase-3, Clone: 31A893, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056709

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056709

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Primary or Secondary Primary
Gene ID (Entrez) 836
Conjugate Unlabeled
Gene Accession No. P42574
Monoclonal or Polyclonal Monoclonal
Formulation PBS containing 0.05% BSA
Cross Reactivity Human
Format Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Clone 31A893
Host Species Mouse
Antigen Caspase-3
Regulatory Status RUO
Quantity 0.1mg
Immunogen A recombinant full-length human Caspase-3 protein was used as the immunogen for this antibody.
Isotype IgG