Please login to your online account to display your discounted pricing

anti-Caspase-3, Clone: 31A893, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056709SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056709T

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Formulation PBS containing 0.05% BSA
Gene Accession No. P42574
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 836
Concentration 0.5mg/mL
Antigen Caspase-3
Format Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.025mg
Immunogen A recombinant full-length human Caspase-3 protein was used as the immunogen for this antibody.
Regulatory Status RUO
Clone 31A893
Isotype IgG
Cross Reactivity Human
Host Species Mouse