Please login to your online account to display your discounted pricing

anti-Caspase-2, Clone: 18E809.3, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056686SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056686T

  • / Each
 

Description & Specifications

Specifications

Antigen Caspase-2
Applications Western Blot
Clone 18E809.3
Conjugate Unlabeled
Cross Reactivity Human
Format Ascites
Formulation PBS containing 0.2% gelatin
Gene Accession No. P42575
Host Species Murine
Immunogen A peptide corresponding to amino acids 400-450 of human Caspase-2 was used as immunogen. GenBank no NP_116764.
Isotype κ
Quantity 0.05mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 835
Test Specificity In HL60, a 48 kDa band is observed. This will cross-react with mouse and rat Caspase-2, based on BLAST. Cross reacts with Human. Predicted to react with Mouse and Rat.
Target Species Human