Please login to your online account to display your discounted pricing

anti-Caspase-2, Clone: 18E809.3, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056686

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056686

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Test Specificity In HL60, a 48 kDa band is observed. This will cross-react with mouse and rat Caspase-2, based on BLAST. Cross reacts with Human. Predicted to react with Mouse and Rat.
Conjugate Unlabeled
Gene ID (Entrez) 835
Gene Accession No. P42575
Monoclonal or Polyclonal Monoclonal
Formulation PBS containing 0.2% gelatin
Applications Western Blotting
Quantity 0.2mL
Cross Reactivity Human
Format Ascites
Host Species Mouse
Regulatory Status RUO
Immunogen A peptide corresponding to amino acids 400-450 of human Caspase-2 was used as immunogen. GenBank no NP_116764.
Isotype κ
Clone 18E809.3
Antigen Caspase-2