Please login to your online account to display your discounted pricing

anti-Cas9, Clone: 7A9-3A3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP236440

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP236440

  • / Each
 

Description & Specifications

Specifications

Antigen Cas9
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blot
Clone 7A9-3A3
Concentration 1.0mg/mL
Conjugate Unlabeled
Cross Reactivity Bacteria
Format Purified
Formulation PBS
Host Species Murine
Immunogen Recombinant Cas9 within the N-terminal region of &italic_begin;Streptococcus pyogene&italic_end;. [UniProt No. Q99ZW2]
Isotype κ
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Cas9 protein from Streptococcus pyogene serotype M1.
Target Species Bacteria