Please login to your online account to display your discounted pricing

anti-Carbonic Anhydrase IX/CA9, Clone: 2D3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP151691

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP151691

  • / Each
 

Description & Specifications

Specifications

Formulation PBS
Gene Accession No. Q16790
Gene ID (Entrez) 768
Primary or Secondary Primary
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Isotype IgG1
Regulatory Status RUO
Conjugate Unlabeled
Concentration 1.0mg/mL
Format Purified
Cross Reactivity Human
Clone 2D3
Immunogen Purified recombinant fragment of human Carbonic Anhydrase IX expressed in &italic_begin;E. coli&italic_end;. [UniProt No. Q16790]
Host Species Mouse
Quantity 0.1mL
Antigen Carbonic Anhydrase IX/CA9