Please login to your online account to display your discounted pricing

anti-Calnexin, Clone: 3H4A7, Novus Biologicals™

$144.63 - $363.90

Specifications

Clone 3H4A7
Host Species Murine
Isotype IgG2b
Immunogen A synthetic peptide (CEAAEERPWLWVVYILTVAL) corresponding to a region of human Calnexin
Antigen Calnexin
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP147546SS

 
novus biologicals canada
NBP147546SS
Unlabeled 0.025mL Each for $144.63

NBP147546

 
novus biologicals canada
NBP147546
Unlabeled 0.1mL Each for $363.90
Description & Specifications

Specifications

Clone 3H4A7
Host Species Murine
Isotype IgG2b
Immunogen A synthetic peptide (CEAAEERPWLWVVYILTVAL) corresponding to a region of human Calnexin
Antigen Calnexin
Monoclonal or Polyclonal Monoclonal
Format Ascites
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Gene Accession No. P27824
Gene ID (Entrez) 821