Please login to your online account to display your discounted pricing

anti-Calcium-sensing R/CaSR, Clone: HL 1499, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1001830

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1001830

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P41180
Primary or Secondary Primary
Concentration 1.0mg/mL
Formulation Tris-glycine, 150mM NaCl
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 846
Immunogen N-terminal
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Radioimmunoassay
Applications Simple Western
Applications Western Blotting
Isotype κ
Clone HL 1499
Quantity 0.1mL
Antigen Calcium-sensing R/CaSR
Host Species Mouse
Format Purified