Please login to your online account to display your discounted pricing

anti-Calcium-sensing R/CaSR, Clone: HL 1499, Novus Biologicals™


Manufacturer: novus biologicals canada  NB1001830SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB1001830SS

  • / Each
 

Description & Specifications

Specifications

Antigen Calcium-sensing R/CaSR
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Radioimmunoassay
Applications Simple Western
Applications Western Blot
Clone HL 1499
Concentration 1.0mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rabbit
Cross Reactivity Rat
Format Purified
Formulation Tris-glycine, 150mM NaCl
Gene Accession No. P41180
Host Species Murine
Immunogen N-terminal
Isotype κ
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 846
Target Species Human
Target Species Murine
Target Species Rabbit
Target Species Rat