Please login to your online account to display your discounted pricing

anti-Calcium-sensing R/CaSR, Clone: 5C10, ADD, Novus Biologicals™

$110.17 - $110.17

Specifications

Clone 5C10
Host Species Murine
Isotype IgG2a
Immunogen Synthetic peptide corresponding to amino acids 214-235 of Human Calcium Sensing Receptor. ADDDYGRPGIEKFREEAEERDI
Antigen Calcium-sensing R/CaSR
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB12019347T

 
novus biologicals canada
NB12019347SS
Unlabeled 0.02mg Each for $110.17
Description & Specifications

Specifications

Clone 5C10
Host Species Murine
Isotype IgG2a
Immunogen Synthetic peptide corresponding to amino acids 214-235 of Human Calcium Sensing Receptor. ADDDYGRPGIEKFREEAEERDI
Antigen Calcium-sensing R/CaSR
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity Has been reported in the literature to work in Mouse (PMID: 21969374).
Gene Accession No. P41180
Gene ID (Entrez) 846
Formulation PBS with 1mg/mL BSA
Concentration 1.0mg/mL