Please login to your online account to display your discounted pricing

anti-Calcium-sensing R/CaSR, Clone: 5C10, ADD, Novus Biologicals™


Manufacturer: novus biologicals canada  NB12019347SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB12019347T

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Regulatory Status RUO
Antigen Calcium-sensing R/CaSR
Host Species Mouse
Immunogen Synthetic peptide corresponding to amino acids 214-235 of Human Calcium Sensing Receptor. ADDDYGRPGIEKFREEAEERDI
Clone 5C10
Quantity 0.02mg
Format Purified
Isotype IgG2a
Formulation PBS with 1mg/mL BSA
Monoclonal or Polyclonal Monoclonal
Test Specificity Has been reported in the literature to work in Mouse (PMID: 21969374).
Gene Accession No. P41180
Primary or Secondary Primary
Gene ID (Entrez) 846
Concentration 1.0mg/mL
Conjugate Unlabeled