Please login to your online account to display your discounted pricing

anti-CA125/MUC16, Clone: X75, Novus Biologicals™


Manufacturer: novus biologicals canada  NB6001468

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB6001468

  • / Each
 

Description & Specifications

Specifications

Format Ion Exchange Chromatography
Applications Sandwich ELISA
Applications Western Blotting
Isotype IgG1
Antigen CA125/MUC16
Immunogen CA-125 purified from human ovarian carcinoma.
Clone X75
Cross Reactivity Human
Host Species Mouse
Quantity 0.1mg
Primary or Secondary Primary
Conjugate Unlabeled
Formulation 10mM Tris, pH 7.5, 0.15M NaCl
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 94025
Test Specificity This specifically reacts with human ovarian carcinoma cells; specifically epitope specificity group B (ISOBM classification). Not yet tested in other species.
Regulatory Status RUO