Please login to your online account to display your discounted pricing

anti-CA125/MUC16, Clone: X75, Novus Biologicals™

$109.00 - $219.00

Specifications

Clone X75
Host Species Murine
Isotype IgG1
Immunogen CA-125 purified from human ovarian carcinoma.
Antigen CA125/MUC16
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB6001468

 
novus biologicals canada
NB6001468
Unlabeled 0.1mg Each for $219.00

NB6001468SS

 
novus biologicals canada
NB6001468SS
Unlabeled 0.02mg Each for $109.00
Description & Specifications

Specifications

Clone X75
Host Species Murine
Isotype IgG1
Immunogen CA-125 purified from human ovarian carcinoma.
Antigen CA125/MUC16
Monoclonal or Polyclonal Monoclonal
Format Ion Exchange Chromatography
Primary or Secondary Primary
Regulatory Status RUO
Applications Sandwich ELISA
Applications Western Blot
Test Specificity This specifically reacts with human ovarian carcinoma cells; specifically epitope specificity group B (ISOBM classification). Not yet tested in other species.
Gene ID (Entrez) 94025
Formulation 10mM Tris, pH 7.5, 0.15M NaCl