Please login to your online account to display your discounted pricing

anti-CA125/MUC16, Clone: X75, Novus Biologicals™


Manufacturer: novus biologicals canada  NB6001468SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB6001468SS

  • / Each
 

Description & Specifications

Specifications

Antigen CA125/MUC16
Applications Sandwich ELISA
Applications Western Blot
Clone X75
Conjugate Unlabeled
Cross Reactivity Human
Format Ion Exchange Chromatography
Formulation 10mM Tris, pH 7.5, 0.15M NaCl
Host Species Murine
Immunogen CA-125 purified from human ovarian carcinoma.
Isotype IgG1
Quantity 0.02mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 94025
Test Specificity This specifically reacts with human ovarian carcinoma cells; specifically epitope specificity group B (ISOBM classification). Not yet tested in other species.
Target Species Human