Please login to your online account to display your discounted pricing

anti-C-Reactive Protein/CRP, Clone: C6, Novus Biologicals™

$219.00 - $329.00

Specifications

Clone C6
Host Species Mouse
Isotype IgG2a
Immunogen Full length native protein (purified) (Human).
Antigen C-Reactive Protein/CRP
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB20044201

 
novus biologicals canada
NB20044201MG
Unlabeled 0.1mg Each for $219.00

NB200442

 
novus biologicals canada
NB200442
Unlabeled 0.5mg Each for $329.00
Description & Specifications

Specifications

Clone C6
Host Species Mouse
Isotype IgG2a
Immunogen Full length native protein (purified) (Human).
Antigen C-Reactive Protein/CRP
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunoassay
Applications Immunohistochemistry
Applications Sandwich ELISA
Test Specificity Human CRP.
Gene Accession No. P02741
Gene ID (Entrez) 1401
Formulation PBS, pH 7.4