Please login to your online account to display your discounted pricing

anti-c-Myc, Clone: MYC699, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232838

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232838

  • / Each
 

Description & Specifications

Specifications

Immunogen A synthetic peptide
Cross Reactivity Human
Cross Reactivity Primate
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Isotype κ
Format Purified
Antigen c-Myc
Quantity 0.5mL
Clone MYC699
Host Species Mouse
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Test Specificity Reacts with Chimpanzee, Gorilla. Mouse (Negative).
Primary or Secondary Primary
Conjugate Unlabeled
Gene ID (Entrez) 4609
Gene Accession No. P01106
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL