Please login to your online account to display your discounted pricing

anti-c-Myc, Clone: MYC699, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328381ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328381M

  • / Each
 

Description & Specifications

Specifications

Immunogen A synthetic peptide
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Format Purified
Host Species Mouse
Clone MYC699
Isotype κ
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Primate
Monoclonal or Polyclonal Monoclonal
Test Specificity Reacts with Chimpanzee, Gorilla. Mouse (Negative).
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL
Gene ID (Entrez) 4609
Gene Accession No. P01106
Primary or Secondary Primary
Antigen c-Myc
Quantity 1mL