Please login to your online account to display your discounted pricing

anti-c-Myc, Clone: MYC275, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232839

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232839

  • / Each
 

Description & Specifications

Specifications

Quantity 0.5mL
Cross Reactivity Human
Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Antigen c-Myc
Isotype κ
Clone MYC275
Immunogen Recombinant human c-myc protein
Regulatory Status RUO
Gene Accession No. P01106
Concentration 0.2mg/mL
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Primary or Secondary Primary
Gene ID (Entrez) 4609
Host Species Mouse
Monoclonal or Polyclonal Monoclonal