Please login to your online account to display your discounted pricing

anti-c-Myc, Clone: MYC275, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone MYC275
Host Species Mouse
Isotype κ
Immunogen Recombinant human c-myc protein
Antigen c-Myc
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232839

 
novus biologicals canada
NBP232839
Unlabeled 0.5mL Each for $321.00

NBP2328391M

 
novus biologicals canada
NBP2328391ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone MYC275
Host Species Mouse
Isotype κ
Immunogen Recombinant human c-myc protein
Antigen c-Myc
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P01106
Gene ID (Entrez) 4609
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL