Please login to your online account to display your discounted pricing

anti-c-Myc, Clone: MYC275, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328391ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328391M

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Recombinant human c-myc protein
Isotype κ
Quantity 1mL
Clone MYC275
Antigen c-Myc
Host Species Mouse
Cross Reactivity Human
Format Purified
Monoclonal or Polyclonal Monoclonal
Gene Accession No. P01106
Primary or Secondary Primary
Gene ID (Entrez) 4609
Concentration 0.2mg/mL
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Regulatory Status RUO