Please login to your online account to display your discounted pricing

anti-c-Myc, Clone: CT14.G4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232960

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232960

  • / Each
 

Description & Specifications

Specifications

Immunogen A synthetic peptide
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Antigen c-Myc
Cross Reactivity Human
Cross Reactivity Primate
Quantity 0.5mL
Isotype κ
Regulatory Status RUO
Clone CT14.G4
Primary or Secondary Primary
Gene ID (Entrez) 4609
Monoclonal or Polyclonal Monoclonal
Test Specificity Reacts with Chimpanzee, Gorilla. Mouse (Negative).
Formulation PBS with 0.05% BSA.
Gene Accession No. P01106
Concentration 0.2mg/mL
Conjugate Unlabeled
Format Purified