Please login to your online account to display your discounted pricing

anti-c-Myc, Clone: CT14.G4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329601ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329601M

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen A synthetic peptide
Cross Reactivity Human
Cross Reactivity Primate
Host Species Mouse
Isotype κ
Antigen c-Myc
Format Purified
Quantity 1mL
Clone CT14.G4
Gene ID (Entrez) 4609
Test Specificity Reacts with Chimpanzee, Gorilla. Mouse (Negative).
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL
Conjugate Unlabeled
Gene Accession No. P01106
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO