Please login to your online account to display your discounted pricing

anti-c-Myc, Clone: 9E11, Novus Biologicals™


Manufacturer: novus biologicals canada  NB200108

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB200108

  • / Each
 

Description & Specifications

Specifications

Test Specificity Human, mouse, Saccharomyces and chicken.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4609
Formulation PBS
Primary or Secondary Primary
Concentration 1.1mg/mL
Gene Accession No. P01106
Applications Chromatin Immunoprecipitation
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Regulatory Status RUO
Quantity 0.1mL
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 408-420 (AEEQKLISEEDL) of human c-Myc
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Yeast
Isotype κ
Antigen c-Myc
Format Purified
Clone 9.00E+11