Please login to your online account to display your discounted pricing

anti-c-Myb, Clone: MYB286, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328371ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328371M

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Immunogen A synthetic peptide
Quantity 1mL
Isotype κ
Regulatory Status RUO
Clone MYB286
Primary or Secondary Primary
Gene ID (Entrez) 4602
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Gene Accession No. P10242
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL
Antigen c-Myb