Please login to your online account to display your discounted pricing

anti-c-Myb, Clone: MYB286, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232837

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232837

  • / Each
 

Description & Specifications

Specifications

Isotype κ
Antigen c-Myb
Format Purified
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.5mL
Immunogen A synthetic peptide
Clone MYB286
Cross Reactivity Human
Primary or Secondary Primary
Gene Accession No. P10242
Monoclonal or Polyclonal Monoclonal
Formulation PBS with 0.05% BSA.
Gene ID (Entrez) 4602
Concentration 0.2mg/mL
Host Species Mouse
Conjugate Unlabeled