Please login to your online account to display your discounted pricing

anti-c-Myb, Clone: MYB286, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone MYB286
Host Species Mouse
Isotype κ
Immunogen A synthetic peptide
Antigen c-Myb
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232837

 
novus biologicals canada
NBP232837
Unlabeled 0.5mL Each for $321.00

NBP2328371M

 
novus biologicals canada
NBP2328371ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone MYB286
Host Species Mouse
Isotype κ
Immunogen A synthetic peptide
Antigen c-Myb
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. P10242
Gene ID (Entrez) 4602
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL