Please login to your online account to display your discounted pricing

anti-BST2, Clone: 4F6, Novus Biologicals™

$109.00 - $259.02

Specifications

Clone 4F6
Host Species Murine
Isotype κ
Immunogen Full length recombinant human BST2 protein [NCBI NP_004326]
Antigen BST2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP229622SS

 
novus biologicals canada
NBP229622SS
Unlabeled 0.025mg Each for $109.00

NBP229622

 
novus biologicals canada
NBP229622
Unlabeled 0.1mg Each for $259.02
Description & Specifications

Specifications

Clone 4F6
Host Species Murine
Isotype κ
Immunogen Full length recombinant human BST2 protein [NCBI NP_004326]
Antigen BST2
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene ID (Entrez) 684
Formulation PBS
Concentration 1.0mg/mL