Please login to your online account to display your discounted pricing

anti-Bromodeoxyuridine/BrdU, Clone: BU20a, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329221ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329221M

  • / Each
 

Description & Specifications

Specifications

Quantity 1mL
Host Species Mouse
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Cross Reactivity All
Isotype IgG1
Format Purified
Immunogen Bromodeoxyuridine (BrdU) conjugated to KLH
Regulatory Status RUO
Clone BU20a
Antigen Bromodeoxyuridine/BrdU
Formulation PBS with 0.05% BSA.
Primary or Secondary Primary
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL